Zinn ≥99,9985% (Metall-Basis), Folie, Puratronic®, Dicke 1.0 mm (0.04 in)

Lieferant: Thermo Fisher Scientific

11457.FG 11457.FY 11457.HL
11457.FGEA 83.2 EUR
11457.FG 11457.FY 11457.HL
Zinn ≥99,9985% (Metall-Basis), Folie, Puratronic®, Dicke 1.0 mm (0.04 in)
Zinn

Purity: 99.9985% CAS No.: 7440-31-5 Physical Form: ≈9.1g/25x50mm Odour: Odourless MDL No.: MFCD00133862

Formel: Sn
Siedepunkt: 2270 °C (1013 hPa)
Schmelzpunkt: 232 °C
Dichte: 7,31 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00133862
CAS-Nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR