Zinn ≥99,998% (Metall-Basis), Folie, Puratronic®, Dicke 0.25 mm (0.01 in)

Lieferant: Thermo Fisher Scientific

11459.FI 11459.GH
11459.FIEA 75.6 EUR
11459.FI 11459.GH
Zinn ≥99,998% (Metall-Basis), Folie, Puratronic®, Dicke 0.25 mm (0.01 in)
Zinn

Purity: 99.998% CAS No.: 7440-31-5 Physical Form: ≈4.6g/50x50mm Odour: Odourless MDL No.: MFCD00133862

Formel: Sn
Siedepunkt: 2270 °C (1013 hPa)
Schmelzpunkt: 232 °C
Dichte: 7,31 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00133862
CAS-Nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR