Silber ≥99,9985% (Metall-Basis), Drahtform, Premion®, Ø 0.25 mm (0.01 in)

Lieferant: Thermo Fisher Scientific

11468.G5 11468.G9 11468.H4
11468.G5EA 93.7 EUR
11468.G5 11468.G9 11468.H4
Silber ≥99,9985% (Metall-Basis), Drahtform, Premion®, Ø 0.25 mm (0.01 in)
Silver

Purity: 99.9985% CAS No.: 7440-22-4 Physical Form: ≈0.51g/m Odour: Characteristic MDL No.: MFCD00003397

Formel: Ag
Siedepunkt: 2212 °C (1013 hPa)
Schmelzpunkt: 962 °C
Dichte: 10,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003397
CAS-Nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR