Silber ≥99,997% (Metall-Basis), Drahtform, Premion®, Ø 0.1 mm (0.004 in)

Lieferant: Thermo Fisher Scientific

11470.G2 11470.G6 11470.H2
11470.G2EA 58.4 EUR
11470.G2 11470.G6 11470.H2
Silber ≥99,997% (Metall-Basis), Drahtform, Premion®, Ø 0.1 mm (0.004 in)
Silver

Purity: 99.997% CAS No.: 7440-22-4 Physical Form: ≈0.08g/m Odour: Characteristic MDL No.: MFCD00003397

Formel: Ag
Siedepunkt: 2212 °C (1013 hPa)
Schmelzpunkt: 962 °C
Dichte: 10,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003397
CAS-Nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR