Antimon(III)fluorid ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Antimontrifluorid

11492.22 11492.36
11492.22EA 60.7 EUR
11492.22 11492.36
Antimon(III)fluorid ≥99%
Antimon(III)fluorid

Purity: 99+% CAS No.: 7783-56-4 Molecular Formula: SbF3 Formula Weight: 178.75 Physical Form: Crystalline Density: 4.379 Melting Point: 292° Boiling Point: 376° Merck Reference: 14,709 UN No.: 2923 MDL No.: MFCD00011218

Formel: F₃Sb
Siedepunkt: 376 °C
Schmelzpunkt: 292 °C
Dichte: 4,379
Lagertemperatur: Raumtemperatur
MDL: MFCD00011218
CAS-Nummer: 7783-56-4
EINECS: 232-009-2
UN: 2923
ADR: 8,III
Merck Index: 14,709

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR