Polishing cloths, Tork
Lieferant: Essity
197270
115-1612EA 81 EUR
115-1612
Polishing cloths, Tork
Wischtücher Reinigungstücher, wiederverwendbar Tücher für sensible Anwendungen
These low-linting cloths made from viscose and polyester are ideal for polishing sensitive surfaces. The soft and flexible cloth releases polishing agents for a professional shine without leaving any traces.
  • Leave no scratches
  • Perfect for working with sensitive surfaces due to low-lint properties
  • Silicone free cloth ensures there are no residues on a surface
  • Folded version: Tork Easy Handling™ for easier carrying, opening, and disposing of packaging
The folded version can be used in the Tork folded wiper/cloth dispenser (W4) that protects the refills from dirt and offers an one-at-a time dispensing to reduce consumption and waste.
The rolls can be used in either the Tork floor or wall stand dispensers (W1) or the Tork Maxi centre-feed (W2) or boxed combi roll dispenser (W3), which are designed for smoother single-hand dispensing.