These economical wipes made of 100% polypropylene in a triple layer construction are impregnated with a 70/30 solution of IPA (Isopropyl Alcohol) and DI water and provide excellent cleaning and disinfection of hard surfaces. They are ideal for cleaning cleanrooms and other areas where contamination control is important.
- Ideal for hard surface disinfection in medical, healthcare and laboratory environments
- Can be used to pre-clean or decontaminate surfaces prior to sterilisation or high level disinfection
- Fabric provides excellent release properties to lay disinfectants on surfaces plus reabsorb dissolved soils
- Available as standard and double-bagged irradiated version
Wipes are effective against gram-positive and gram-negative bacteria and most viruses and fungi, such as MRSA, SARS, VRE, Salmonella, HIV and TB.
Weight: 18 g/m²
Verpackung: 200 wipes per cleanroom compatible plastic tub. Irradiated version is double bagged and supplied with a certificate of irradiation
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904