Enghalsflaschen, viereckig, mit Schraubverschluss, Azlon®
Lieferant: SciLabware
BSC0500 BSC1000 BSC2000 BSC0250
115-1790EA 226 EUR
115-1790 115-1791 115-1792 115-1789
Enghalsflaschen, viereckig, mit Schraubverschluss, Azlon®
Flaschen Kunststoffflaschen
Polycarbonate, highly transparent.
  • Leakproof
  • Suitable for repeated sterilisation by autoclaving at +121 °C
  • Practical square design for economic use of space
  • Heavy-duty and durable
Ideal for the preparation and storage of biological media, reagents and aqueous solutions. With linerless polypropylene cap.