TX1009 - AlphaWipe®, Reinraumtuch, 9 x9 (23cm x 23cm), Polyester-Gestrick 1 * 150 ST
Lieferant: Texwipe
TX 1009 AlphaWipe PES suitable for ISO class 5-6 cleanrooms, constructed from 100% polyester filament, cleanroom laundred cut edge, basis weight approx. 155g/m² basis weight approx. 155g/m² 150 wipers/bag (2 inner bags of 75 wipers) 1 * 150 items