Reinraum-Eingangsmatten, BioClean Supatac™
Lieferant: Nitritex
STB1845 STB3645 STW2645 STW3645 STW1845
115-3050EA 123 EUR
115-3050 115-3051 115-3053 115-3054 115-1359
Reinraum-Eingangsmatten, BioClean Supatac™
Fußmatten Klebematten
Supatac™ Access Mats are made from LDPE with Polyacrylate adhesive and consist of 30 sheets. They prevent contamination and particulate matter from entering controlled and critical working environments. Manufactured with an anti-bacterial finish, they represent the most advanced technology for significantly reducing the level of traffic-borne contaminants.
  • Suitable for ISO Cleanroom Class 5
  • Uniform thickness and width without pinholes and High Durability Standard Adhesive on entire surface
  • 30 sheets: each sheet coated with a unique adhesive
  • Convenient
More Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable