Palladium ≥99,9% (Metall-Basis), Folie, Dicke 0.5 mm

Lieferant: Thermo Fisher Scientific

11514.FF 11514.FI
11514.FFEA 1211 EUR
11514.FF 11514.FI
Palladium ≥99,9% (Metall-Basis), Folie, Dicke 0.5 mm
Palladium

Purity: 99.9% CAS No.: 7440-05-3 Physical Form: ≈3.8g/25x25mm Odour: Odourless MDL No.: MFCD00011167

Formel: Pd
Molecular Weight: 106.420 g/mol
Dichte: 12.02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011167
CAS-Nummer: 7440-05-3
EINECS: 231-115-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR