Iridium ≥99,7% (Metall-Basis), Folie, Dicke 1.0 mm (0.04 in)
Lieferant: Thermo Fisher Scientific
11525.FF
11525.FFEA
8178
EUR
11525.FF
Iridium ≥99,7% (Metall-Basis), Folie, Dicke 1.0 mm (0.04 in)
Iridium
Purity: 99.7+%
CAS No.: 7439-88-5
Physical Form: ≈14.2g/25x25mm
MDL No.: MFCD00011062
Formel:
Ir Siedepunkt: 5300 °C (1013 hPa) Schmelzpunkt: 2450 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011062 CAS-Nummer: 7439-88-5 EINECS: 231-095-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...