Zinn(II)fluorid ≥97,5%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97.5%
CAS No.: 7783-47-3
Molecular Formula: F2Sn
Formula Weight: 156.69
Physical Form: Crystalline
Density: 4.57
Appearance: White to off-white
Odour: Odourless
Melting Point: 219°
Boiling Point: 850°
Merck Reference: 14,8784
UN No.: 3260
MDL No.: MFCD00042540
Formel:
SnF₂ Siedepunkt: 850 °C Schmelzpunkt: 219 °C Dichte: 4,57 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042540 CAS-Nummer: 7783-47-3 EINECS: 231-999-3 UN: 3260 ADR: 8,II Merck Index: 14,8784 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...