Wolframdisilicid ≥99,5% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

11551.18
11551.18EA 166 EUR
11551.18
Wolframdisilicid ≥99,5% (Metall-Basis)
Tungsten disilicide

Purity: 99.5% CAS No.: 12039-88-2 Molecular Formula: WSi2 Formula Weight: 240.02 Physical Form: -325 Mesh Powder Odour: Odourless MDL No.: MFCD00049704

Formel: Si₂W
Schmelzpunkt: 2165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049704
CAS-Nummer: 12039-88-2
EINECS: 234-909-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR