Kaliumfluorid Dihydrat ≥98,5%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
11555.14
11555.30
11555.A1
11555.14EA
18.8
EUR
11555.14
11555.30
11555.A1
Kaliumfluorid Dihydrat ≥98,5%
Kaliumfluorid Dihydrat
Purity: 98.5+%
CAS No.: 13455-21-5
Molecular Formula: KF.2H2O
Formula Weight: 94.13 (58.10anhy)
Physical Form: Crystalline
Density: 2.5
Refractive Index: 1.352
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 41°
Solubility: Freely soluble in water
Merck Reference: 14,7632
UN No.: 1812
MDL No.: MFCD00149929
Formel:
FK·2H₂O Siedepunkt: 1505 °C (1013 hPa) Schmelzpunkt: 41 °C Dichte: 2,5 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149929 CAS-Nummer: 13455-21-5 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III Merck Index: 14,7632 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...