Zinkacetat Dihydrat 98,0-101,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
11559.30
11559.A1
11559.A7
11559.30EA
49.4
EUR
11559.30
11559.A1
11559.A7
Zinkacetat Dihydrat 98,0-101,0% ACS
Zinkacetat Dihydrat
Purity: 98.0-101.0%
CAS No.: 5970-45-6
Molecular Formula: C4H10O6Zn
Formula Weight: 219.50 (183.47anhy)
Physical Form: Crystalline
Density: 1.735
Appearance: White
Odour: Weak, characteristic
pH: 6-7
Melting Point: 237°
Solubility: Soluble in water, alcohol
Beilstein Reference: 3732513
Merck Reference: 14,10128
UN No.: 3077
MDL No.: MFCD00066961
Formel:
(H₃CCOO)₂Zn·2H₂O Schmelzpunkt: 237 °C Dichte: 1,735 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066961 CAS-Nummer: 5970-45-6 EINECS: 209-170-2 UN: 3077 ADR: 9,III Merck Index: 14,10128 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...