Chrom(III)nitrat Nonahydrat ≥98,5%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chromtrinitrat Nonahydrat

11565.14 11565.30 11565.A1
11565.14EA 34.4 EUR
11565.14 11565.30 11565.A1
Chrom(III)nitrat Nonahydrat ≥98,5%
Chrom(III)nitrat Nonahydrat

Purity: 98.5% CAS No.: 7789-02-8 Molecular Formula: Cr(NO3)3.9H2O Formula Weight: 400.14 (238.01anhy) Physical Form: Crystalline Appearance: Purple Odour: Odourless Melting Point: 60° Solubility: Soluble in water, alcohol Merck Reference: 14,2226 UN No.: 2720 MDL No.: MFCD00149659


Catalyst, corrosion inhibitor, in textile printing

Formel: CrN₃O₉·9H₂O
Schmelzpunkt: 60 °C
Dichte: 1,807 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149659
CAS-Nummer: 7789-02-8
EINECS: 236-921-1
UN: 2720
ADR: 5.1,III
Merck Index: 14,2226

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR