Ammoniumfluorid ≥98,0% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Fluoroammonium

11568.14 11568.30 11568.A1 11568.D9
11568.14EA 21.5 EUR
11568.14 11568.30 11568.A1 11568.D9
Ammoniumfluorid ≥98,0% ACS
Ammonium fluoride

Purity: 98.0% min CAS No.: 12125-01-8 Molecular Formula: NH4F Formula Weight: 37.04 Physical Form: Crystalline Density: 1.01 Solubility: Soluble in cold water (0°C). Decomposes in hot water to NH3 and ammonium bifluoride. Slightly soluble in alcohol Merck Reference: 14,522 UN No.: 2505 MDL No.: MFCD00011423


For etching glass, in printing textile, for preservation of wood

Formel: NH₄F
Schmelzpunkt: 160 °C
Dichte: 1,01
Lagertemperatur: Raumtemperatur
MDL: MFCD00011423
CAS-Nummer: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 14,522

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR