Rhodium ≥99,8% (Metall-Basis), Drahtform, Ø 0.5 mm (0.02 in)

Lieferant: Thermo Fisher Scientific

11576.BS 11576.BR 11576.BV 11576.CB
11576.BSEA 627 EUR
11576.BS 11576.BR 11576.BV 11576.CB
Rhodium ≥99,8% (Metall-Basis), Drahtform, Ø 0.5 mm (0.02 in)
Rhodium
Formel: Rh
Molecular Weight: 102.9 g/mol
Siedepunkt: 3400…3700 °C (1013 hPa)
Schmelzpunkt: 1960
Dichte: 12.41 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011201
CAS-Nummer: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR