Natriumdihydrogenphosphat Monohydrat 98,0-102,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Natriumphosphat monobasisch Monohydrat
11591.30
11591.A1
11591.A7
11591.30EA
41.3
EUR
11591.30
11591.A1
11591.A7
Natriumdihydrogenphosphat Monohydrat 98,0-102,0% ACS
Natriumdihydrogenphosphat Monohydrat
Purity: 98.0-102.0%
CAS No.: 10049-21-5
Molecular Formula: NaH2PO4.H2O
Formula Weight: 137.99 (119.98anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Merck Reference: 14,8660
MDL No.: MFCD00149208
Formel:
NaH₂PO₄·H₂O Schmelzpunkt: 100 °C (-H₂O) Dichte: 2,04 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149208 CAS-Nummer: 10049-21-5 EINECS: 231-449-2 Merck Index: 14,8660 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...