di-Natriumhydrogenphosphat Heptahydrat 98,0-102,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98.0-102.0%
CAS No.: 7782-85-6
Molecular Formula: Na2HPO4.7H2O
Formula Weight: 268.07 (141.98anhy)
Physical Form: Crystalline
Density: 1.7
Appearance: White
Odour: Odourless
Melting Point: 48.1°-5H2O
Merck Reference: 14,8659
MDL No.: MFCD00149180
Formel:
HNa₂O₄P·7H₂O Schmelzpunkt: 48,1 °C (-5H₂O) Dichte: 1,7 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149180 CAS-Nummer: 7782-85-6 EINECS: 231-448-7 Merck Index: 14,8659 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...