Ammoniumacetat ≥97,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Essigsäure Ammoniumsalz
Purity: 97.0% min
CAS No.: 631-61-8
Molecular Formula: CH3COONH4
Formula Weight: 77.08
Flash Point: 136°(277°F)
Physical Form: Crystalline
Density: 1.17
Sensitivity: Hygroscopic
Appearance: White
Odour: Ammonia-like
Melting Point: ca 114° dec.
Solubility: Soluble in cold water, approximately 148g/100ml. Decomposes in hot water. Soluble in methanol approximately 7.9g/100ml
Beilstein Reference: 4186741
Merck Reference: 14,495
MDL No.: MFCD00013066
Reagent in analytical chemistry, drugs, textile dying, preserving meats, foam rubbers, vinyl plastics
Formel:
H₃CCOONH₄ Schmelzpunkt: ∼114 °C (Zersetzung) Dichte: 1,17 Flammpunkt: 136 °C (277 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013066 CAS-Nummer: 631-61-8 EINECS: 211-162-9 Merck Index: 14,495 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...