Standard fire fighting equipment signs
Lieferant: Brady
816888 830823 817675 817678 816880 816883 817039 817042 817198 817834 817357 817360 830796
BRDY816888EA 10.53 EUR
BRDY816888 116-0556 116-0544 116-0545 116-0546 116-0547 116-0548 116-0549 116-0550 116-0552 116-0554 116-0555 116-0558
Standard fire fighting equipment signs
Warn- und Verbotszeichen
A range of signs to clearly mark the location of fire extinguishers and other fire fighting equipment as well as fire alarm points.
  • Laminated polyester
  • Resistance to most chemical products and solvents
  • Resistance against extreme temperatures
  • Good weatherability and UV resistance
  • Excellent adhesion to low surface energy surfaces
  • Thickness: 0,1 mm
  • Different sizes
Bestellen
TECHNISCHE DATEN
Environmentally Preferable