Prohibition signs, access roads and entrances
Lieferant: KROSCHKE
Z3059
116-1160EA 207 EUR
116-1160
Prohibition signs, access roads and entrances
Warn- und Verbotszeichen
These prohibition signs are made of plastic.
  • Temperature resistant from –20 to 70 °C
  • Resistant to moisture, fire and weak acids
Ideal for indoor and outdoor use.