Bismut(III)oxid ≥99,5% (durch EDTA Titration), BAKER ANALYZED® für elektronische Keramik, J.T.Baker®
Lieferant: Avantor
Synonyme:
Dibismuttrioxid
1160-08EA
0
EUR
1160-08
Bismut(III)oxid ≥99,5% (durch EDTA Titration), BAKER ANALYZED® für elektronische Keramik, J.T.Baker®
Bismut(III)oxid
Formel:
BiO₃ Molecular Weight: 465,96 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003462 CAS-Nummer: 1304-76-3 |
Spezifikation Testergebnisse
For Microelectronic Use | — |
Assay (as Bi₂O₃) (by EDTA titrn) | ≥99.5 % |
Chloride (Cl) | ≤0.001 % |
Nitrate (NO₃) | ≤0.005 % |
Total Sulfur (as SO₄) | ≤0.002 % |
Aluminum (Al) | ≤0.005 % |
Calcium (Ca) | ≤0.005 % |
Copper (Cu) | ≤0.001 % |
Iron (Fe) | ≤0.003 % |
Lead (Pb) | ≤0.005 % |
Silicon (Si) | ≤0.005 % |
Sodium (Na) | ≤0.001 % |
Average Particle Diameter, µm (APD) | ≤3 |
Specific Surface Area, m²/g, For Information Only | For Information Only |
Bulk Density (g/cc), For Information Only | For Information Only |
Mesh (Wet Screen Analysis) - On U.S. No. 325 Sieve, For Information Only | For Information Only |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...