Natriumperchlorat, wasserfrei 98,0-102,0% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

11623.22 11623.36
11623.22EA 48.9 EUR
11623.22 11623.36
Natriumperchlorat, wasserfrei 98,0-102,0% ACS
Sodium perchlorate

Purity: 98.0-102.0% CAS No.: 7601-89-0 Molecular Formula: NaClO4 Formula Weight: 122.46 Physical Form: Crystalline Density: 2.50 Melting Point: 482° dec. Merck Reference: 14,8653 UN No.: 1502 MDL No.: MFCD00003480

Formel: ClNaO₄
Dichte: 2,50
Lagertemperatur: Raumtemperatur
MDL: MFCD00003480
CAS-Nummer: 7601-89-0
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 14,8653

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR