Iron(II) perchlorate hydrate, Reagenzqualität
Lieferant: Thermo Fisher Scientific
11646.22
11646.36
11646.22EA
44.6
EUR
11646.22
11646.36
Iron(II) perchlorate hydrate, Reagenzqualität
Iron(II) perchlorate hydrate
CAS No.: 335159-18-7
Molecular Formula: Fe(ClO4)2.xH2O
Formula Weight: 254.75(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point:
Formel:
Fe(ClO₄)₂·xH₂O |
MDL:
MFCD00150234 CAS-Nummer: 335159-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...