Kupfer(II)perchlorat Hexahydrat, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Kupferdiperchlorat Hexahydrat
11651.18
11651.30
11651.18EA
55.5
EUR
11651.18
11651.30
Kupfer(II)perchlorat Hexahydrat, Reagenzqualität
Kupfer(II)perchlorat Hexahydrat
CAS No.: 10294-46-9
Molecular Formula: Cu(ClO4)2.6H2O
Formula Weight: 370.53 (262.44anhy)
Physical Form: Crystalline
Density: 2.23
Refractive Index: 1.505
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 82°
Solubility: Soluble in water, methanol, ethanol, acetic acid, acetic anhydride, and acetone
Merck Reference: 14,2647
UN No.: 1481
MDL No.: MFCD00149666
Formel:
Cl₂CuO₈·6H₂O Schmelzpunkt: 82 °C Dichte: 2,23 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149666 CAS-Nummer: 10294-46-9 EINECS: 237-391-4 UN: 1481 ADR: 5.1,II Merck Index: 14,2647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...