Cobalt(II)perchlorat Hexahydrat, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Cobaltdiperchlorat Hexahydrat
11652.09
11652.18
11652.30
11652.09EA
45.2
EUR
11652.09
11652.18
11652.30
Cobalt(II)perchlorat Hexahydrat, Reagenzqualität
Cobalt(II)perchlorat Hexahydrat
CAS No.: 13478-33-6
Molecular Formula: Co(ClO4)2.6H2O
Formula Weight: 366.74 (257.83anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Odour: Odourless
UN No.: 1481
MDL No.: MFCD00150207
Formel:
Cl₂CoO₈·6H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150207 CAS-Nummer: 13478-33-6 EINECS: 236-653-5 UN: 1481 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...