Lysing Matrix B, MP Biomedicals
Lieferant: MP Biomedicals
116540425
116540425.EA 270 EUR
116540425.
Lysing Matrix B, MP Biomedicals
Homogenisatoren Tischhomogenisatoren
Lysing Matrix B tubes are used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores
  • Impact resistant tube containing 0.1mm silica spheres
  • Can be used as an additive for disruption by sonication
  • Use for gram-positive and gram-negative bacteria
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.