Calciumperchlorat Hydrat (12,2 - 13,5% Ca), Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
11655.30
11655.30EA
66.9
EUR
11655.30
Calciumperchlorat Hydrat (12,2 - 13,5% Ca), Reagenzqualität
Calciumperchlorat Hydrat
CAS No.: 13477-36-6
Molecular Formula: Ca(ClO4)2.xH2O
Formula Weight: 238.98(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
UN No.: 1455
MDL No.: MFCD00015971
Formel:
Ca(ClO₄)₂·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015971 CAS-Nummer: 13477-36-6 EINECS: 236-768-0 UN: 1455 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...