Natriumsulfid Hydrat, tech.

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Schwefelnatrium Hydrat, Sulfigran Hydrat, Dinatriumsulfid Hydrat, Natriummonosulfid Hydrat

11664.36 11664.A3
11664.36EA 51 EUR
11664.36 11664.A3
Natriumsulfid Hydrat, tech.
Natriumsulfid Hydrat

Sodium sulfide hydrate is used as an oxygen scavenger in water treatment, bleaching agent in textile industry, desulfurising and as a dechlorinating agent and in leather industry, sulfonation and sulfomethylation agent in chemical industry.


It is involved in the production of rubber chemicals and sulfur dyes. It is also used to prepare 3-nitroaniline from 1,3-dinitrobenzene and thioethers from alkyl halide. It plays an important role in other applications such as ore flotation, oil recovery, making dyes, and detergent. In addition, it is used in pulp and paper industry in the Kraft process.

Formel: Na₂S·xH₂O
Siedepunkt: ∼920 °C (1013 hPa)
Schmelzpunkt: ∼ 50 °C (Zersetzung)
Dichte: 1,427
Lagertemperatur: Raumtemperatur
MDL: MFCD00149183
CAS-Nummer: 27610-45-3
EINECS: 215-211-5
UN: 1849
ADR: 8,II
REACH: 01-2119513694-38
Merck Index: 14,8681

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR