Adenosine 3ʹ5ʹ-cyclic monophosphateN⁶-benzoyl-sodium salt ≥98% (durch HPLC), Millipore®
Lieferant: Merck Millipore Calbiochem
116802-10
116802-10EA
214
EUR
116802-10
Adenosine 3ʹ5ʹ-cyclic monophosphateN⁶-benzoyl-sodium salt ≥98% (durch HPLC), Millipore®
Adenosine 3ʹ5ʹ-cyclic monophosphateN⁶-benzoyl-sodium salt
A potent cell-permeable activator of cAMP-dependent protein kinase (PKA). Due to its considerably increased lipophilicity, it has sufficient membrane permeability for many biological systems.
- Activates both type 1 and type 2 PKA
- Binds to the cAMP binding site 2 on the regulatory subunit of PKA
- It is only slowly metabolized by cyclic nucleotide-dependent phosphodiesterases
Formel:
C₁₇H₁₅N₅O₇P·Na Lagertemperatur: Gefrierschrank |
CAS-Nummer:
30275-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...