Antimon(III)bromid ≥99,995% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Antimontribromid
11686.09
11686.09EA
115
EUR
11686.09
Antimon(III)bromid ≥99,995% (Metall-Basis)
Antimon(III)bromid
Purity: 99.995%
CAS No.: 7789-61-9
Molecular Formula: SbBr3
Formula Weight: 361.48
Physical Form: Crystalline
Density: 4.148
Refractive Index: 1.74
Sensitivity: Moisture Sensitive
Odour: Characteristic
Melting Point: 96.6°
Boiling Point: 280°
Merck Reference: 14,706
UN No.: 3260
MDL No.: MFCD00016317
Formel:
Br₃Sb Molecular Weight: 361,47 g/mol Siedepunkt: 280 °C Schmelzpunkt: 96,6 °C Dichte: 4,148 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016317 CAS-Nummer: 7789-61-9 EINECS: 232-179-8 UN: 3260 ADR: 8,II Merck Index: 14,706 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...