Ruthenium 5% auf Aluminiumoxid, Pulver

Lieferant: Thermo Fisher Scientific

11749.06 11749.14 11749.22
11749.06EA 61.9 EUR
11749.06 11749.14 11749.22
Ruthenium 5% auf Aluminiumoxid, Pulver
Ruthenium

CAS No.: 7440-18-8 Physical Form: Powder Appearance: Greyish white Odour: Odourless MDL No.: MFCD00011207


Applications: Hydrogenation of aliphatic carbonyls and aromatic rings, especially bulky molecules

Formel: Ru
Lagertemperatur: Raumtemperatur
MDL: MFCD00011207
CAS-Nummer: 7440-18-8
EINECS: 231-127-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR