Eyewash station, HypaClens
Lieferant: SAFETY FIRST AID GROUP
E481 E404
118-0118EA 92.4 EUR
118-0118 118-0114
Eyewash station, HypaClens
Erste-Hilfe-Ausstattung Augenduschen
This eye care station with mirror includes two HypaClens eyewash bottles (500 ml) filled with 0,9% sterile saline solution. Ideal for irrigating eyes and general wound washing applications.
  • Robust, wall mountable eyewash station
  • Integrated mirror is ideal for self-administration of eyewash
  • Clear instruction panel indicates correct use of eye wash
  • Highly visible design
By twisting off the top the sterile contents may be applied drop by drop or in a continuous gentle stream by squeezing the bottle, a stronger jet may be delivered by breaking off theMore Product Information