Ruthenium(IV)oxid Hydrat (≥54% Ru)

Lieferant: Thermo Fisher Scientific
Achtung

11803.06 11803.03 11803.14
11803.06EA 446 EUR
11803.06 11803.03 11803.14
Ruthenium(IV)oxid Hydrat (≥54% Ru)
Ruthenium(IV)oxid Hydrat

CAS No.: 32740-79-7 Molecular Formula: O2Ru.xH2O Formula Weight: 133.07(anhy) Physical Form: Powder Appearance: Black Odour: Odourless UN No.: 1479 MDL No.: MFCD00149846


Oxidation

Formel: O₂Ru·H₂O
Schmelzpunkt: 1200 °C (Subl.)
Dichte: 6,97 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149846
CAS-Nummer: 32740-79-7
EINECS: 234-840-6
ADR: ,

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR