Ruthenium(III) bromide (≥29% Ru)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
11807.06
11807.06EA
414
EUR
11807.06
Ruthenium(III) bromide (≥29% Ru)
Ruthenium(III) bromide
CAS No.: 14014-88-1
Molecular Formula: RuBr3
Formula Weight: 340.78
Physical Form: Powder
Density: 5.30
Sensitivity: Hygroscopic
Odour: Characteristic
UN No.: UN3260
MDL No.: MFCD00016314
Formel:
RuBr₃ Dichte: 5,30 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016314 CAS-Nummer: 14014-88-1 EINECS: 237-829-4 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...