Ammoniumheptamolybdat Tetrahydrat 81-83% (MoO₃-Basis) ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
11831.22
11831.18
11831.36
11831.A3
11831.22EA
57.1
EUR
11831.22
11831.18
11831.36
11831.A3
Ammoniumheptamolybdat Tetrahydrat 81-83% (MoO₃-Basis) ACS
Ammoniumheptamolybdat Tetrahydrat
Purity: 81-83%
CAS No.: 12054-85-2
Molecular Formula: H24Mo7N6O24.4H2O
Formula Weight: 1235.86 (1163.79anhy)
Physical Form: Powder
Density: 2.498
Appearance: White to pale green
Melting Point: ca 190° dec.
Solubility: Soluble in water (1:2 ratio). Insoluble in alcohol
Merck Reference: 14,533
MDL No.: MFCD00167059
In photography, analytical lab reagent for phosphate, lead, arsenate determination
Formel:
(NH₄)₆Mo₇O₂₄·4H₂O Schmelzpunkt: ∼ 190 °C (Zersetzung) Dichte: 2,498 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00167059 CAS-Nummer: 12054-85-2 EINECS: 234-722-4 Merck Index: 14,533 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...