Thallium(I)chlorid ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.999%
CAS No.: 7791-12-0
Molecular Formula: TlCl
Formula Weight: 239.82
Physical Form: Powder
Density: 7.0
Refractive Index: 2.247
Sensitivity: Air Sensitive
Melting Point: 430°
Merck Reference: 14,9260
UN No.: 1707
MDL No.: MFCD00011274
Formel:
ClTl Siedepunkt: 720 °C (1013 hPa) Schmelzpunkt: 430 °C Dichte: 7,0 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011274 CAS-Nummer: 7791-12-0 EINECS: 232-241-4 UN: 1707 ADR: 6.1,II Merck Index: 14,9260 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...