Bismut(III)chloridoxid ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bismutoxychlorid
11843.09
11843.18
11843.09EA
79.9
EUR
11843.09
11843.18
Bismut(III)chloridoxid ≥99,999% (Metall-Basis)
Bismuth chloride oxide
Purity: 99.999%
CAS No.: 7787-59-9
Molecular Formula: BiClO
Formula Weight: 260.43
Physical Form: Powder
Density: 7.72
Refractive Index: 2.15
Appearance: Very pale brown
Odour: Odourless
Melting Point: dec.
Merck Reference: 14,1263
MDL No.: MFCD00010895
Formel:
BiClO Schmelzpunkt: Zersetzung Dichte: 7,72 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010895 CAS-Nummer: 7787-59-9 EINECS: 232-122-7 Merck Index: 14,1263 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...