Indium(III)iodid, wasserfrei ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

11854.04 11854.09 11854.18
11854.04EA 102 EUR
11854.04 11854.09 11854.18
Indium(III)iodid, wasserfrei ≥99,999% (Metall-Basis)
Indium(III)iodid

Purity: 99.999% CAS No.: 13510-35-5 Molecular Formula: I3In Formula Weight: 495.53 Physical Form: Crystalline Density: 4.69 Appearance: Yellow to brown Melting Point: 210° UN No.: 3260 MDL No.: MFCD00016151

Formel: I₃In
Siedepunkt: 500 °C (1013 hPa)
Schmelzpunkt: 210 °C
Dichte: 4,69
Lagertemperatur: Raumtemperatur
MDL: MFCD00016151
CAS-Nummer: 13510-35-5
EINECS: 236-839-6
UN: 3260
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR