Indium(III)chlorid, wasserfrei ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

11856.03 11856.09 11856.18 11856.30
11856.03EA 53.9 EUR
11856.03 11856.09 11856.18 11856.30
Indium(III)chlorid, wasserfrei ≥99,999% (Metall-Basis)
Indium(III)chlorid

Purity: 99.999% CAS No.: 10025-82-8 Molecular Formula: InCl3 Formula Weight: 221.18 Physical Form: Crystalline Density: 3.460 Appearance: White Odour: Odourless Melting Point: 586° Boiling Point: 800° Merck Reference: 14,4958 UN No.: 3260 MDL No.: MFCD00011058

Formel: Cl₃In
Siedepunkt: 800 °C
Schmelzpunkt: 586 °C
Dichte: 3,460
Lagertemperatur: Raumtemperatur
MDL: MFCD00011058
CAS-Nummer: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8,II
Merck Index: 14,4958

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR