Cadmium(II)carbonat ≥99,99% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

11864.14
11864.14EA 85.1 EUR
11864.14
Cadmium(II)carbonat ≥99,99% (Metall-Basis)
Cadmium(II)carbonat

Purity: 99.99% CAS No.: 513-78-0 Molecular Formula: CdCO3 Formula Weight: 172.42 Physical Form: Powder/Lumps Density: 4.26 Melting Point: 500° dec. Merck Reference: 14,1616 UN No.: 2570 MDL No.: MFCD00010918

Formel: CCdO₃
Molecular Weight: 172,42 g/mol
Schmelzpunkt: 500 °C (Zers.)
Dichte: 4,26
Lagertemperatur: Raumtemperatur
MDL: MFCD00010918
CAS-Nummer: 513-78-0
EINECS: 208-168-9
UN: 2570
ADR: 6.1,III
Merck Index: 14,1616

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR