Sodium hexachloroiridate(III) hydrate (≥31,5% Ir)

Lieferant: Thermo Fisher Scientific
Achtung

11889.03 11889.06
11889.03EA 261 EUR
11889.03 11889.06
Sodium hexachloroiridate(III) hydrate (≥31,5% Ir)
Sodium hexachloroiridate(III) hydrate

Sodium hexachloroiridate(III) hydrate, is used as a reagent for HPLC, including analyses of peptides and proteins.


Molecular Formula: Na3IrCl6.xH2O Formula Weight: 473.89(anhy) Physical Form: Crystalline Sensitivity: Moisture Sensitive MDL No.: MFCD00003508

Formel: Na₃IrCl₆·xH₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00003508
CAS-Nummer: 123334-23-6
EINECS: 239-795-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR