Silber(I)thiocyanat
Lieferant: Thermo Scientific
|
Achtung
|
11897.09
11897.09EA
322
EUR
11897.09
Silber(I)thiocyanat
Silber(I)thiocyanat
CAS No.: 1701-93-5
Molecular Formula: CAgNS
Formula Weight: 165.95
Physical Form: Crystalline
Sensitivity: Light Sensitive
Odour: Odourless
UN No.: 3077
MDL No.: MFCD00003416
Preparation of nonmetallic thiocyanates, analytical reagent, in photographic emulsions, as an organic intermediate
Formel:
AgSCN Molecular Weight: 165,95 g/mol Schmelzpunkt: Zersetzung Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003416 CAS-Nummer: 1701-93-5 EINECS: 216-934-9 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...