These safety drum vents provide automatic pressure relief for drums between 0,20 bar but less than 0,55 bar. Depending on the model, vacuum relief is either automatic (on horizontally installed vents) or manual (on vertically installed vents).
Vents can prevent escape of flammable vapour in closed position