Drum locks
Lieferant: JUSTRITE MANUFACTURING
08510
119-0235EA 187 EUR
119-0235
Drum locks
Sicherheitsverriegelungen
Drum locks easily install around the drum bung flange over the original bung.
  • Made of corrosion resistant
  • Non-sparking zinc die cast
  • Locks install in seconds to secure contents and reduce theft, pilfering, and sabotage
There is no contact with the drum contents, the potential for content contamination is eliminated and making them suitable for chemical and food processing applications.