EDTA (Ethylendiamintetraessigsäure) ≥99,4% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.4+%
CAS No.: 60-00-4
Molecular Formula: C10H16N2O8
Formula Weight: 292.24
Physical Form: Powder
Appearance: White
Melting Point: ca 245° dec.
Beilstein Reference: 1716295
Merck Reference: 14,3517
MDL No.: MFCD00003541
Chelating agent
Formel:
C₁₀H₁₆N₂O₈ Siedepunkt: 614 °C (1013 hPa) Dichte: 0,86 g/cm³ (20 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003541 CAS-Nummer: 60-00-4 EINECS: 200-449-4 REACH: 01-2119486399-18 Merck Index: 14,3517 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...