Blei ≥99,998% (Metall-Basis), Folie, Puratronic®, Dicke 0.5 mm (0.02 in)

Lieferant: Thermo Fisher Scientific
Gefahr

12049.FF 12049.FI
12049.FFEA 92.9 EUR
12049.FF 12049.FI
Blei ≥99,998% (Metall-Basis), Folie, Puratronic®, Dicke 0.5 mm (0.02 in)
Lead

Purity: 99.998% CAS No.: 7439-92-1 Physical Form: ≈3.5g/25x25mm Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

Formel: Pb
Siedepunkt: 1740 °C (1013 hPa)
Schmelzpunkt: 327 °C
Dichte: 11,3437 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00134050
CAS-Nummer: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR